missing translation for 'onlineSavingsMsg'
Learn More

CEACAM6/CD66c Antibody (B6.2) - BSA Free, Novus Biologicals™

Product Code. 18368662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368662 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368662 Supplier Novus Biologicals Supplier No. NBP309125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM6/CD66c
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone B6.2
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 3-12 ug/ml, Immunoprecipitation
Formulation PBS
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Host Species Mouse
Immunogen Extracts from human breast carcinoma cells
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4680
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.