missing translation for 'onlineSavingsMsg'
Learn More

CEACAM6/CD66c Antibody (1H7-4B), DyLight 680, Novus Biologicals™

Product Code. 18424773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424773 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424773 Supplier Novus Biologicals Supplier No. NBP254626FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM6/CD66c
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 1H7-4B
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Host Species Mouse
Immunogen Recombinant human CEACAM6/CD66c extracellular domain.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4680
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.