missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Specifications
Specifications
| Antigen | CEACAM6/CD66c |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | 1H7-4B |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen |
| Host Species | Mouse |
| Immunogen | Recombinant human CEACAM6/CD66c extracellular domain. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?