missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5, Mouse anti-Human, Clone: COL-1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15172.100ug
This item is not returnable.
View return policy
Specifications
| CEACAM5 | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS. | |
| CD66e/CEA/DKFZp781M2392 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| COL-1 | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| carcinoembryonic antigen-related cell adhesion molecule 5 | |
| CEACAM5 | |
| Human colon carcinoma extract. | |
| RUO | |
| 1048/634 | |
| Store at 4°C. For long term storage store at -20°C to -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction