missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Mouse anti-Human, Clone: COL-1, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00 - £405.00
Specifications
| Antigen | CEACAM5/CD66e |
|---|---|
| Clone | COL-1 |
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18371852
|
Novus Biologicals
NBP3-05766-100UG |
100 μg |
£405.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670068
|
Novus Biologicals
NBP3-05766-20ug |
20 μg |
£215.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CEACAM5/CD66e | |
| 0.2 mg/mL | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 1048 | |
| Primary | |
| Store at 4°C. |
| COL-1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| biosimilar, biosimilars, Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Human colon carcinoma extract | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title