missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (SPM506), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47975AF350
This item is not returnable.
View return policy
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| CEACAM5/CD66e | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Human colon carcinoma extract (Uniprot: P06731) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Immunology | |
| 1048 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| SPM506 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction