missing translation for 'onlineSavingsMsg'
Learn More

CEACAM5/CD66e Antibody (C66/1292), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18431244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431244 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431244 Supplier Novus Biologicals Supplier No. NBP254412AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM5/CD66e
Applications Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone C66/1292
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Host Species Mouse
Immunogen Recombinant full-length human CEACAM5/CD66e protein (Uniprot: P06731)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.