missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | CEACAM5/CD66e |
| Applications | ELISA, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | C66/1291 |
| Conjugate | DyLight 405 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human CEACAM5/CD66e protein (Uniprot: P06731) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?