missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (487609), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB41281I
This item is not returnable.
View return policy
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| CEACAM5/CD66e | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 487609 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Mouse myeloma cell line NS0-derived recombinant human CEACAM-5/CD66e, Lys35-Ala685, Accession # ABM87752 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Immunology | |
| 1048 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction