missing translation for 'onlineSavingsMsg'
Learn More

CEACAM5/CD66e Antibody (3E10-3), DyLight 550, Novus Biologicals™

Product Code. 18433493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433493 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433493 Supplier Novus Biologicals Supplier No. NBP254628R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM5/CD66e
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Clone 3E10-3
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Host Species Mouse
Immunogen Recombinant human CEACAM5/CD66e
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.