missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (308/3-3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £319.00
Specifications
| Antigen | CEACAM5/CD66e |
|---|---|
| Clone | 308/3-3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA, Immunoprecipitation 1:10 - 1:500, Sandwich ELISA 1:100 - 1:2000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Sandwich ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609167
|
Novus Biologicals
NBP2-54624-0.025mg |
0.025 mg |
£169.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619157
|
Novus Biologicals
NBP2-54624 |
0.1 mg |
£319.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Sandwich ELISA, CyTOFSpecifications
| CEACAM5/CD66e | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Sandwich ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Immunology | |
| PBS | |
| 1048 | |
| IgG1 κ | |
| Protein A purified |
| 308/3-3 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA, Immunoprecipitation 1:10 - 1:500, Sandwich ELISA 1:100 - 1:2000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Recombinant soluble human CEACAM3-Fc produced in HEK293 cells | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title