missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM4 Monoclonal Antibody (822608)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561877
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
CEACAM4 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 4) is a Protein Coding gene. An important paralog of this gene is CEACAM3.
Specifications
| CEACAM4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75871 | |
| CEACAM4 | |
| E. coli-derived recombinant human CEACAM-4, Phe36-Val140. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 822608 | |
| Unconjugated | |
| CEACAM4 | |
| carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 10; carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; C-CAM4 protein; Ceacam10; CEACAM4; CEA-related cell adhesion molecule 10; CGM7; CGM7_HUMAN; NCA; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction