missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM3 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148808
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| CEACAM3 | |
| Polyclonal | |
| Unconjugated | |
| CEACAM3 | |
| carcinoembryonic antigen CGM1; Carcinoembryonic antigen gene family (CGM1); carcinoembryonic antigen gene family 4; carcinoembryonic antigen gene family member 1; Carcinoembryonic antigen gene family member 4 / (carcinoembryonic antigene-related protein); carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen-related cell adhesion molecule 3; CD66d; CD66d antigen; CEA; cea12; CEACAM3; ceacam3 {ECO:0000312; Ceacam5; Cear; Cgm1; Cgm4; CGM4AA; EG384557; nonspecific cross-reacting antigen; pregnancy-specific glycoprotein 24; Psg24; RATCEAA; RATCGM4AA; RGD:2333}; W264; W282 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 1084 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P40198 | |
| CEACAM3 | |
| Mouse myeloma cell line NS0-derived recombinant human CEACAM-3/CD66d isoform 1, Lys35-Gly155. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction