missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM3/CD66d Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92411-0.1ml
This item is not returnable.
View return policy
Description
CEACAM3/CD66d Polyclonal antibody specifically detects CEACAM3/CD66d in Human, Mouse samples. It is validated for Western Blot
Specifications
| CEACAM3/CD66d | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-135 of human CEACAM3/CD66d (NP_001806.2). MGPPSASPHRECIPWQGLLLTASLLNFWNPPTTAKLTIESMPLSVAEGKEVLLLVHNLPQHLFGYSWYKGERVDGNSLIVGYVIGTQQATPGAAYSGRETIYTNASLLIQNVTQNDIGFYTLQVIKSDLVNEEAT | |
| 0.1 mL | |
| CD Markers, Immunology, Innate Immunity | |
| 1084 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction