missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570882
This item is not returnable.
View return policy
Description
CEACAM20 Polyclonal Antibody for Western Blot
Together with the tyrosine-protein kinase SYK, enhances production of the cytokine CXCL8/IL-8 via the NFKB pathway and may thus have a role in the intestinal immune response.
Specifications
| CEACAM20 | |
| Polyclonal | |
| Unconjugated | |
| CEACAM20 | |
| 9130012D09Rik; carcinoembryonic antigen related cell adhesion molecule 20; carcinoembryonic antigen-related cell adhesion molecule 20; Ceacam20; CEA-related cell adhesion molecule 20; GPAD9366; UNQ9366; UNQ9366/PRO34155 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 125931 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UY09 | |
| CEACAM20 | |
| synthetic peptide directed towards the C-terminal region of human CEACAM20. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction