missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553171
This item is not returnable.
View return policy
Description
Immunogen sequence: TRNWRGQSHR LPAPRGQGSL SILCSAVSPV PSVTPSTWMA TTEKPELGPA HDAGDNNIYE VMPSPVLLVS PISDTRSINP ARPLPTPPHL QAEPENHQYQ QDLLNPDPA Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 38%.
The specific function of the protein remains unknown.
Specifications
| CEACAM19 | |
| Polyclonal | |
| Unconjugated | |
| CEACAM19 | |
| C130022P09Rik; carcinoembryonic antigen related cell adhesion molecule 19; carcinoembryonic antigen-like 1; Carcinoembryonic antigen-related cell adhesion molecule 19; CEACAM19; CEACM19; CEAL1; CEA-related cell adhesion molecule 19; UNQ2973/PRO7436 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z692 | |
| CEACAM19 | |
| Recombinant protein corresponding to Human CEACAM19. Recombinant protein control fragment (Product #RP-90570). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction