missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEACAM19 Monoclonal Antibody (1042108)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA562023

Product Code. 30534148

  • £324.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM19
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q7Z692
CEACAM19
Chinese hamster ovary cell line CHO-derived recombinant human CEACAM-19, Met1-Gly157.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry
1042108
Unconjugated
CEACAM19
C130022P09Rik; carcinoembryonic antigen related cell adhesion molecule 19; carcinoembryonic antigen-like 1; Carcinoembryonic antigen-related cell adhesion molecule 19; CEACAM19; CEACM19; CEAL1; CEA-related cell adhesion molecule 19; UNQ2973/PRO7436
Mouse
Protein A/G
RUO
56971
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.