missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM19 Antibody (Z5P2D9*A9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53124AF532
This item is not returnable.
View return policy
Description
CEACAM19 Monoclonal specifically detects CEACAM19 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CEACAM19 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CEACAM19 | |
| Peptide sequence - RSINPARPLP | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| Z5P2D9*A9 | |
| Western Blot, ELISA | |
| carcinoembryonic antigen-related cell adhesion molecule 19 | |
| Mouse | |
| Protein A purified | |
| Apoptosis, Cancer | |
| 56971 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction