missing translation for 'onlineSavingsMsg'
Learn More

CEACAM1 Mouse anti-Human, PE, Clone: CLB-gran/10, Invitrogen™

Product Code. 15205874
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity unitSize
15205874 100 Tests 100 tests
1 options
This item is not returnable. View return policy

Product Code. 15205874

Brand: Invitrogen A15787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM1
Applications Flow Cytometry
Classification Monoclonal
Clone CLB-gran/10
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene Alias BGP, BGP1, BGPI
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 634
Target Species Human
Content And Storage Store at 2 - 8 °C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.