missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEACAM1/CEACAM3/CEACAM6 Monoclonal Antibody (YTH71.3)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA517003

Product Code. 15357900

  • £504.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody does not react with mouse. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

This gene encodes a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of all variants has not been defined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM1/CEACAM3/CEACAM6
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P13688, P40198, P40199
CEACAM1, CEACAM3, CEACAM6
Human lymphocytes.
200 μg
Primary
Human, Rhesus Macaque
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
YTH71.3
Unconjugated
CEACAM1
antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen CGM1; carcinoembryonic antigen CGM6; carcinoembryonic antigen gene family member 1; carcinoembryonic antigen gene family member 6; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen related cell adhesion molecule 8; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); carcinoembryonic antigen-related cell adhesion molecule 8; CD66a; CD66a antigen; CD66b; CD66c; CD66D; CD66d antigen; CD67; CD67 antigen; CEA; CEA cell adhesion molecule 1; CEA cell adhesion molecule 3; CEA cell adhesion molecule 6; CEACAM1; CEACAM3; CEACAM6; CEACAM8; CEAL; CGM1; CGM6; Cluster of Differentiation 66c; NCA; NCA-95; Non-specific crossreacting antigen; nonspecific cross-reacting antigen; non-specific cross-reacting antigen NCA-95; normal cross-reacting antigen; pan CEACAM; W264; W282
Rat
Protein G
RUO
106994775, 1084, 4680, 634, 707824, 708200
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.