missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM1/CD66a Antibody (F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59673
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Binds specific to the N-domain of rat CEACAM1. Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| F7 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CEACAM1 | |
| The immunogen for this antibody is proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 634 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction