missing translation for 'onlineSavingsMsg'
Learn More

CEA Antibody (PARLAM 4), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18422727 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422727 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422727 Supplier Novus Biologicals Supplier No. NBP197718MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEA Monoclonal antibody specifically detects CEA in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEA
Applications Flow Cytometry
Classification Monoclonal
Clone PARLAM 4
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Carcino Embryonic Antigen, Carcinoembryonic antigen
Host Species Mouse
Immunogen This antibody is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 Ag 14 Mouse myeloma cells with spleen cells from a BABL/c Mouse immunized with isolated Human CEA. The immunogen has been isolated from Human colonic carcinoma cells.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.