missing translation for 'onlineSavingsMsg'
Learn More

CE3F4, MedChemExpress

Product Code. p-200101609
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16444208

Brand: Medchem Express HY108539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CE3F4 is a selective antagonist of exchange protein directly activated by cAMP (Epac1), with IC50s of 10.7 μM and 66 μM for Epac1 and Epac2(B), respectively.

CE3F4 is a selective antagonist of exchange protein directly activated by cAMP (Epac1), with IC50s of 10.7 μM and 66 μM for Epac1 and Epac2(B), respectively.

Specifications

Chemical Name or Material CE3F4
CAS 143703-25-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H10Br2FNO
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (142.45 mM; Need ultrasonic)
SMILES O=CN1C(C)CCC2=C1C=C(Br)C(F)=C2Br
Molecular Weight (g/mol) 351.01
Formula Weight 351.01
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.