missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CE3F4, MedChemExpress
Description
CE3F4 is a selective antagonist of exchange protein directly activated by cAMP (Epac1), with IC50s of 10.7 μM and 66 μM for Epac1 and Epac2(B), respectively.
Specifications
Specifications
| Chemical Name or Material | CE3F4 |
| CAS | 143703-25-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H10Br2FNO |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (142.45 mM; Need ultrasonic) |
| SMILES | O=CN1C(C)CCC2=C1C=C(Br)C(F)=C2Br |
| Molecular Weight (g/mol) | 351.01 |
| Formula Weight | 351.01 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?