Learn More
Vileda Professional™ CE Tank Cleaner
Specialty above-the-floor tools are ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting
Brand:  Vileda Professional™ 162234
Additional Details : Weight : 0.34000kg
Product Code. 17862466
  • £253.30 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • Ideal for inner and outer tank walls
  • Manufactured of electro-polished stainless steel
  • Completely autoclavable
  • Frame is pre-bent to 9 in. radius but is easily adjusted, and can be flat to use on isolators
  • 4 degree of freedom to ensure good contact with the surface being cleaned
  • Easy to use with handle for tight spaces and can be used with telescopic handle for hard to reach areas
Specifications
Specifications
Tank Cleaning Tool
Non-sterile
9 in.
Inner and Outer Tanks Walls
Silver
Autoclavable
1
Electropolished Stainless Steel