missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ CE Multi-Surface Cleaner™

Product Code. 17882466
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
17882466 Autoclavable Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17882466 Supplier Vileda Professional™ Supplier No. 162236
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleaning tool is designed to clean pipes, hoods and other hard to reach or uneven surfaces. Ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting.

  • Designed to clean pipes, hoods and other hard to reach or uneven surfaces
  • Manufactured of high performance plastics and stainless steel
  • Completely autoclavable and rust free
  • Versatile
  • Easily bendable to give the tool different shapes to effectively clean any surface

Specifications

Product Type Multi-Surface Cleaning Tool
Color Blue
Sterility Non-sterile
Autoclavable Autoclavable
Size 23 in.
Quantity 1
For Use With (Application) Pipes, Hoods, Hard to reach Areas
Material Stainless Steel, Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.