Learn More
Vileda Professional™ CE MicroIntensive Duo™
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 25
Description
- Independent lab study, conducted by BMA Labs, resulting in bacteria reduction of 99.99%
- 4-layer construction - featuring Polyester/Microfiber embossed cleaning side
- Pre-laundered for single-use
- Gamma and autoclave compatible
- Extremely low particle shedding
- Double-sided cleaning with color -stitched side indicator
- Corner drainage openings for cleaner rinses
- Finger loop for touchless on/off of cleaning sides
Specifications
Specifications
| Product Type | Mop |
| Color | White |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Material | Polyester Microfiber |