missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDYL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550010
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CDYL2 protein.
CDYL2 (Chromodomain Y Like 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includemethylated histone binding. An important paralog of this gene isCDYL.
Specifications
| CDYL2 | |
| Polyclonal | |
| Unconjugated | |
| CDYL2 | |
| 1700029M19Rik; 4930453I21Rik; Cdyl2; CDY-like 2; chromodomain protein, Y chromosome-like 2; chromodomain protein, Y-like 2; chromodomain Y-like protein 2; Gm20194; PCCP1; prostate cancer candidate protein 1, PCCP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124359 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N8U2 | |
| CDYL2 | |
| Synthesized peptide derived from N-terminal of humanCDYL2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction