missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDY1B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05641-100ul
This item is not returnable.
View return policy
Description
CDY1B Polyclonal antibody specifically detects CDY1B in Mouse, Rat samples. It is validated for Western Blot
Specifications
| CDY1B | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CDY, chromodomain protein, Y chromosome, 1, centromeric, chromodomain protein, Y-linked, 1B, testis-specific chromodomain protein on Y, centromeric, testis-specific chromodomain protein Y 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human CDY1B (NP_001003895.1). EKQKKLTWTTTSRIFSNNARRRTSRSTKANYSKNSPKTPVTDKHHRSKNRKLFAASKNVRRKAASILSDTKNMEIINSTIETLAPDSPFDHKTVSGFQKLEKLDPIAADQQDTVVFKVTEGKLLRDPLSRPGAEQTGIQNKTQIHPLMSQMSGSVTASMATGSATRKGIVVLIDPLAANGTTDMHTSVPRVKGGQRNITDDSRDQPFIKKMHFTIRLTESASTYRDIVVKKEDGFTQIVLSTRSTEKNALNTEVIKEIVNA | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 253175 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction