missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDY1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £359.00
Specifications
| Antigen | CDY1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621177
|
Novus Biologicals
NBP3-05663-100ul |
100 μg |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689968
|
Novus Biologicals
NBP3-05663-20ul |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDY1 Polyclonal antibody specifically detects CDY1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CDY1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CDY, CDY1A, Chromodomain Protein, Y Chromosome, 1, Chromodomain Protein, Y-Linked, 1, Chromodomain Y-Linked 1, EC 2.3.1.48 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human CDY1 (NP_004671.1). MASQEFEVEAIVDKRQDKNGNTQYLVRWKGYDKQDDTWEPEQHLMNCEKCVHDFNRRQTEKQKKLTWTTTSRIFSNNARRRTSRSTKANYSKNSPKTPVTDKHHRSKNRKLFAASKNVRRKAASILSDTKNMEIINSTIETLAPDSPFDHKTVSGFQKLEKLDPIAADQQDTVVFKVTEGKLLRDPLSRPGAEQTGIQNKTQIHPLMSQM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9085 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title