missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22943-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.Specifications
| CDX4 | |
| Polyclonal | |
| Unconjugated | |
| CDX4 | |
| caudal type homeobox 4, Caudal type homeobox protein 4, CDX4, Homeobox protein CDX 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1046, 12592 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14627, Q07424 | |
| CDX4 | |
| CDX4 Fusion Protein Ag19078 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction