missing translation for 'onlineSavingsMsg'
Learn More

CDX4 Antibody (2H7), Novus Biologicals™

Product Code. 18361608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361608 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361608 Supplier Novus Biologicals Supplier No. H00001046M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDX4 Monoclonal antibody specifically detects CDX4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2H7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005184
Gene Alias caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4
Host Species Mouse
Immunogen CDX4 (NP_005184, 202 a.a. ~ 284 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RKSELAVNLGLSERQVKIWFQNRRAKERKMIKKKISQFENSGGSVQSDSDSISPGELPNTFFTTPSAVRGFQPIEIQQVIVSE*
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1046
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.