missing translation for 'onlineSavingsMsg'
Learn More

CDX2 Rabbit Recombinant Antibody, Proteintech

Product Code. 30235256 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235256 20 μL 20µL
30236139 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235256 Supplier Proteintech Supplier No. 826591RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The CDX2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CDX2. This antibody recognizes human, mouse, rat antigen. The CDX2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.

Specifications

Antigen CDX2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
Clone 3J1
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols CDX-3; CDX3
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 33-40 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.