missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523397
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen for the GATA-4 antibody is 93% homologous in Rhesus Macaque, 87% homologous in Rat, 82% in Mouse, and 81% homologous in Canine.
The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| CDX2 | |
| Polyclonal | |
| Unconjugated | |
| CDX2 | |
| caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100051406, 1045, 12591, 618679, 66019 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| P43241, Q99626 | |
| CDX2 | |
| A portion of amino acids 1-60 of human CDX2/CDX3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Horse, Marsupial | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction