missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Mouse anti-Human, Clone-CDX2/1690, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54472-100UG
This item is not returnable.
View return policy
Description
CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.
Specifications
| CDX2 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Mouse | |
| 40 kDa | |
| 100 μg | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 1045 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| CDX2/1690 | |
| Unconjugated | |
| PBS with No Preservative | |
| CDX2 | |
| Recombinant human CDX2 protein fragment (aa150-249) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction