missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDX2 Monoclonal Antibody (ZC007)
GREENER_CHOICE

Product Code. 10017432
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10017432 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10017432 Supplier Invitrogen™ Supplier No. 397800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CDX2 Monoclonal Antibody for Western Blot, IHC (P), ELISA

The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ZC007
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CDX2
Gene Accession No. Q99626
Gene Alias caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor
Gene Symbols CDX2
Host Species Mouse
Immunogen Recombinant full-length human CDX2 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.