missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (rCDX2/9241), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | CDX2 |
|---|---|
| Clone | rCDX2/9241 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626204
|
Novus Biologicals
NBP3-23671-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640214
|
Novus Biologicals
NBP3-23671-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CDX2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 1045 | |
| IgG2b κ | |
| Protein A or G purified |
| rCDX2/9241 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Recombinant fragment (around aa100-300) of human CDX2 protein (exact sequences are proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title