missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (PCRP-CDX2-1A3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75759JF646
This item is not returnable.
View return policy
Description
CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CDX2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDX2 | |
| Recombinant human full-length CDX2 protein (Uniprot: Q99626) | |
| 0.1 mL | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| The specificity of this monoclonal antibody to its intended target was validated by HuProtTMArray, containing more than 19,000, full-length human proteins. The intestine-specific transcription factors CDX1 and CDX2 are important for directing intestinal development, differentiation, proliferation and maintenance of the intestinal phenotype. CDX2 protein expression has been seen in GI carcinomas. Anti-CDX2 has been useful to establish GI origin of metastatic adenocarcinomas and carcinoids and is especially useful to distinguish metastatic colorectal adenocarcinoma from lung adenocarcinoma. However, mucinous carcinomas of the ovary also express CDX2 protein. It limits the usefulness of this marker in the distinction of metastatic colorectal adenocarcinoma from mucinous carcinoma of the ovary. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PCRP-CDX2-1A3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction