Learn More
CDX2 Antibody (CL12967) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18571AF405
Product Code. 30294221
  • £582.00 / 0.10mL
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDX2
Monoclonal
Alexa Fluor 405
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
This antibody was developed using a synthetic peptide derived from Q99626, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
1045
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL12967
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified