missing translation for 'onlineSavingsMsg'
Learn More

CDX2 Antibody (CDX2/1690), DyLight 755, Novus Biologicals™

Product Code. 18425293 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425293 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425293 Supplier Novus Biologicals Supplier No. NBP254472IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Classification Monoclonal
Clone CDX2/1690
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Host Species Mouse
Immunogen Recombinant fragment (around aa150-249) of human CDX2 protein (exact sequence is proprietary) (Uniprot: Q99626)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.