missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdx1 Rabbit anti-Human, Clone: 8F6T7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Cdx1 |
|---|---|
| Clone | 8F6T7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308596
|
Novus Biologicals
NBP3-16822-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18337746
|
Novus Biologicals
NBP3-16822-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdx1 Monoclonal antibody specifically detects Cdx1 in Human samples. It is validated for Western BlotSpecifications
| Cdx1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1044 | |
| IgG | |
| Affinity purified |
| 8F6T7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cdx1 (P47902). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title