missing translation for 'onlineSavingsMsg'
Learn More

CDw17 Antibody (HO18.3G-6.F5) - Azide and BSA Free, Novus Biologicals™

Product Code. 18386094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386094 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386094 Supplier Novus Biologicals Supplier No. NBP311614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDw17 Monoclonal antibody specifically detects CDw17 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDw17
Applications Flow Cytometry, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone HO18.3G-6.F5
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Gene Alias Lactosylceramide
Host Species Mouse
Immunogen beta-2 Microglobulin associated proteins from a detergent lysate of human PBL
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.