missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542861
This item is not returnable.
View return policy
Description
Peptide sequence: PATPPATPPA ASPSALKGVS QDLLERIRAK EAQKQLAQMT RCPEQEQRLQ Sequence homology: Cow: 93%; Dog: 77%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Yeast: 85%.
KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.
Specifications
| CDT1 | |
| Polyclonal | |
| Unconjugated | |
| CDT1 | |
| 2610318F11Rik; AW545653; C76791; CDT1; chromatin licensing and DNA replication factor 1; DNA replication factor Cdt1; Double parked homolog; Double parked, Drosophila, homolog of; DUP; Retroviral insertion site 2 protein; retroviral integration site 1; retroviral integration site 2; RIS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 292071, 67177, 81620 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R4E9, Q9H211 | |
| CDT1 | |
| Synthetic peptide directed towards the C-terminal of human CDT1 (aa 400-449). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction