missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDT1 Antibody (SR3533), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | CDT1 |
|---|---|
| Clone | SR3533 |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041592
|
Novus Biologicals™
NBP3-27779-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041563
|
Novus Biologicals™
NBP3-27779-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDT1 Monoclonal antibody specifically detects CDT1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDT1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| chromatin licensing and DNA replication factor 1, DNA replication factor Cdt1, Double parked homolog, Double parked, Drosophila, homolog of, DUPRIS2 | |
| A synthesized peptide derived from human CDT1 (Uniprot #: Q9H211) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3533 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 81620 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title