missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDT1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2567F
This item is not returnable.
View return policy
Description
CDT1 Polyclonal specifically detects CDT1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CDT1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| chromatin licensing and DNA replication factor 1, DNA replication factor Cdt1, Double parked homolog, Double parked, Drosophila, homolog of, DUPRIS2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 81620 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CDT1 | |
| The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 546) of human Chromatin Licensing and DNA Replication Factor 1 using the numbering given in entry NP_112190.1 (GeneID 81620). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction