missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDSN Polyclonal Antibody

Product Code. 15528262
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15528262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528262 Supplier Invitrogen™ Supplier No. PA547768

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDSN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CDSN
Gene Accession No. Q15517
Gene Alias AI747712; CDSN; Corneodesmosin; D6S586E; differentiated keratinocyte S protein; HTSS; HTSS1; HYPT2; LOC682408; PSS; PSS1; S; S protein; similar to corneodesmosin
Gene Symbols CDSN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Corneodesmosin Lys33-Pro529 (Ser153 del).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1041
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.