missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00008760-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MTELRQRVAHEPVAPPEDKESESEAKVDGETASDSESRAESAPLPVSADDTPEVLNRALSNLSSRWKSpecifications
| CDS2 | |
| Polyclonal | |
| 50 % glycerol | |
| NM_003818 | |
| CDS2 | |
| CDS2 (NP_003809, 1 a.a. ∼ 67 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 8760 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2 | |
| FLJ38111 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction