missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92756-0.02ml
This item is not returnable.
View return policy
Description
CDS2 Polyclonal antibody specifically detects CDS2 in Rat samples. It is validated for Western Blot
Specifications
| CDS2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| CDP-DAG synthase 2, CDP-DG synthase 2, CDP-DG synthetase 2, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2, CDP-diacylglycerol synthase 2, CDP-diglyceride diphosphorylase 2, CDP-diglyceride pyrophosphorylase 2, CDP-diglyceride synthase 2, CDP-diglyceride synthetase 2, CDS 2, CTP:phosphatidate cytidylyltransferase 2, EC 2.7.7, EC 2.7.7.41, FLJ38111, phosphatidate cytidylyltransferase 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDS2 (NP_003809.1). MTELRQRVAHEPVAPPEDKESESEAKVDGETASDSESRAESAPLPVSADDTPEVLNRALSNLSSRWKNWWVRGILTLAMIAFFFIIIYLGPMVLMIIVMC | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 8760 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction