missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDS1 Monoclonal antibody specifically detects CDS1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, KnockDown
Specifications
Specifications
| Antigen | CDS1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | 2D10 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Knockdown Validated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_001254 |
| Gene Alias | CDP-DAG synthase 1, CDP-DG synthase 1, CDP-DG synthetase 1, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 1, CDP-diacylglycerol synthase 1, CDP-diglyceride pyrophosphorylase 1, CDP-diglyceride synthase 1, CDP-diglyceride synthetase 1, CDS, CDS 1, CTP:phosphatidate cytidylyltransferase 1, EC 2.7.7.41, phosphatidate cytidylyltransferase 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?