missing translation for 'onlineSavingsMsg'
Learn More

CDR2L Mouse anti-Human, Mouse, Rat, Clone: 2E12D10, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen CDR2L
Clone 2E12D10
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808995
View Documents
Proteintech
66791-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16898985
View Documents
Proteintech
66791-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerebellar degeneration-related protein 2-like (CDR2L), also named as HUMPPA or Paraneoplastic 62 kDa antigen, is a 465 amino acid protein, which belongs to the CDR2 family. CDR2L contains three potential coiled-coil regions. The functions of protein is so far not known. CDR2L is expressed in the brain and localizes in the cytoplasm. The predicted MW of this protein is 53 kDa.
Specifications

Specifications

CDR2L
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CDR2L, HUMPPA, Paraneoplastic 62 kDa antigen
CDR2L
IgG1
Protein G
Antibody
2E12D10
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Rat, Mouse
A2A6T1, Q86X02
237988, 30850, 360656
CDR2L Fusion Protein Ag6139
Primary
-20°C
CDR2L
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.